Assay Details
Target Gene Details
Entrez Gene ID: | 9590 |
Gene Name: | A-kinase anchoring protein 12 |
Gene Aliases: |
AKAP250, SSeCKS |
Location: |
Chr.6:151239815-151358559 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKAP12 | NM_005100.3 | NP_005091.2 | ||
XM_017011517.1 | XP_016867006.1 | |||
AB210003.1 | ||||
AF001504.1 | AAB58938.1 | |||
AK307578.1 | ||||
BC047728.1 | AAH47728.1 | |||
U81607.1 | AAC51366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1021285 | Chr.6:150819721 - 151343862 on Build GRCh38 | Gain | LOC102723831 MTHFD1L AKAP12 PLEKHG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |