Assay Details
Target Gene Details
Entrez Gene ID: | 55914 |
Gene Name: | erbb2 interacting protein |
Gene Aliases: |
ERBB2IP, HEL-S-78, LAP2 |
Location: |
Chr.5:65926554-66081024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERBIN | NM_001006600.2 | NP_001006600.1 | ||
NM_001253697.1 | NP_001240626.1 | |||
NM_001253698.1 | NP_001240627.1 | |||
NM_001253699.1 | NP_001240628.1 | |||
NM_001253701.1 | NP_001240630.1 | |||
NM_018695.3 | NP_061165.1 | |||
XM_005248554.2 | XP_005248611.1 | |||
XM_005248555.3 | XP_005248612.1 | |||
XM_006714660.3 | XP_006714723.1 | |||
XM_017009636.1 | XP_016865125.1 | |||
XM_017009637.1 | XP_016865126.1 | |||
AF263744.1 | AAF77048.1 | |||
AF276423.1 | AAK69431.1 | |||
AK295708.1 | ||||
AK304693.1 | ||||
BC050692.1 | AAH50692.1 | |||
BC126464.1 | ||||
BC144075.1 | ||||
DA723889.1 | ||||
DR000131.1 | ||||
EU668330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894205 | Chr.5:65896048 - 65955310 on Build GRCh38 | Gain | LOC100303749 ERBIN |
nsv830327 | Chr.5:65910646 - 66032995 on Build GRCh38 | Gain | LOC100303749 ERBIN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |