Assay Details
Target Gene Details
Entrez Gene ID: | 100129792 |
Gene Name: | coiled-coil domain containing 152 |
Gene Aliases: |
CH5400 |
Location: |
Chr.5:42756818-42802437 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC152 | NM_001134848.1 | NP_001128320.1 | ||
AK127368.1 | ||||
AK303469.1 | ||||
BC039102.1 | AAH39102.1 | |||
DB450032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5637n100 | Chr.5:42609988 - 42844089 on Build GRCh38 | Gain | GHR CCDC152 SEPP1 |
nsv523033 | Chr.5:42751935 - 42919583 on Build GRCh38 | Gain | CCDC152 SEPP1 |
nsv597886 | Chr.5:42610307 - 42854306 on Build GRCh38 | Gain | GHR CCDC152 SEPP1 |
nsv597925 | Chr.5:42728729 - 43237086 on Build GRCh38 | Gain | LOC100506639 FLJ32255 ZNF131 LOC648987 CCDC152 ANXA2R LOC100132356 SEPP1 NIM1K LOC153684 |