Assay Details
Target Gene Details
Entrez Gene ID: | 8310 |
Gene Name: | acyl-CoA oxidase 3, pristanoyl |
Gene Aliases: |
- |
Location: |
Chr.4:8366263-8440725 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOX3 | NM_001101667.1 | NP_001095137.1 | ||
NM_003501.2 | NP_003492.2 | |||
XM_005248011.4 | XP_005248068.1 | |||
XM_005248013.3 | XP_005248070.1 | |||
XM_011513565.2 | XP_011511867.1 | |||
XM_011513566.1 | XP_011511868.1 | |||
AK225080.1 | ||||
AK313242.1 | ||||
BC017053.1 | AAH17053.1 | |||
Y11411.1 | CAA72214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593637 | Chr.4:8352102 - 8388919 on Build GRCh38 | Loss |
![]() |
nsv461220 | Chr.4:8371217 - 8456667 on Build GRCh38 | Loss |
![]() ![]() |
nsv1002794 | Chr.4:8108356 - 8489207 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv1005399 | Chr.4:8385554 - 8396256 on Build GRCh38 | Deletion |
![]() |
nsv1009011 | Chr.4:8223942 - 8423104 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)