Assay Details
Target Gene Details
Entrez Gene ID: | 2261 |
Gene Name: | fibroblast growth factor receptor 3 |
Gene Aliases: |
ACH, CD333, CEK2, HSFGFR3EX, JTK4 |
Location: |
Chr.4:1793299-1808872 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGFR3 | NM_000142.4 | NP_000133.1 | ||
NM_001163213.1 | NP_001156685.1 | |||
NM_022965.3 | NP_075254.1 | |||
XM_006713868.1 | XP_006713931.1 | |||
XM_006713869.1 | XP_006713932.1 | |||
XM_006713870.1 | XP_006713933.1 | |||
XM_006713871.1 | XP_006713934.1 | |||
XM_006713872.1 | XP_006713935.1 | |||
XM_006713873.1 | XP_006713936.1 | |||
XM_011513420.1 | XP_011511722.1 | |||
XM_011513422.1 | XP_011511724.1 | |||
AB209441.1 | BAD92678.1 | |||
AF245114.1 | AAF63380.1 | |||
DR003490.1 | ||||
HM244683.1 | ||||
KJ534834.1 | ||||
KJ534974.1 | ||||
KJ535004.1 | ||||
M58051.1 | AAA52450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428434 | Chr.4:1623665 - 1813048 on Build GRCh38 | Gain | FGFR3 FAM53A SLBP TMEM129 TACC3 LETM1 |
nsv949756 | Chr.4:1710274 - 1817773 on Build GRCh38 | Deletion | FGFR3 SLBP TMEM129 TACC3 LETM1 |
dgv8901n54 | Chr.4:1644637 - 1864631 on Build GRCh38 | Loss | FGFR3 FAM53A SLBP TMEM129 TACC3 LETM1 |
nsv508989 | Chr.4:1623823 - 1846590 on Build GRCh38 | Insertion | FGFR3 FAM53A SLBP TMEM129 TACC3 LETM1 |
nsv829836 | Chr.4:1775678 - 1901525 on Build GRCh38 | Loss | FGFR3 WHSC1 LETM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |