Assay Details
Target Gene Details
Entrez Gene ID: | 30061 |
Gene Name: | solute carrier family 40 member 1 |
Gene Aliases: |
FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3 |
Location: |
Chr.2:189560590-189580811 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC40A1 | NM_014585.5 | NP_055400.1 | ||
XM_005246505.1 | XP_005246562.1 | |||
XM_017003938.1 | XP_016859427.1 | |||
AF215636.1 | AAF80986.1 | |||
AF226614.1 | AAF36697.1 | |||
AF231121.1 | AAF44330.1 | |||
AK223236.1 | BAD96956.1 | |||
AK292146.1 | ||||
AK314827.1 | ||||
AL136944.1 | CAB66878.1 | |||
BC035893.1 | AAH35893.1 | |||
BC037733.1 | AAH37733.1 | |||
CR533564.1 | CAG38595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954448 | Chr.2:189550075 - 189580174 on Build GRCh38 | Duplication | SLC40A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |