Assay Details
Target Gene Details
Entrez Gene ID: | 114803 |
Gene Name: | Myb like, SWIRM and MPN domains 1 |
Gene Aliases: |
2A-DUB, 2ADUB |
Location: |
Chr.1:58654739-58700091 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYSM1 | NM_001085487.2 | NP_001078956.1 | ||
XM_006710314.3 | XP_006710377.1 | |||
XM_011540573.2 | XP_011538875.1 | |||
XM_011540574.2 | XP_011538876.1 | |||
XM_017000195.1 | XP_016855684.1 | |||
XM_017000196.1 | XP_016855685.1 | |||
XM_017000197.1 | XP_016855686.1 | |||
AB067502.1 | BAB67808.1 | |||
AK126835.1 | ||||
BX648168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003930 | Chr.1:58464772 - 58691366 on Build GRCh38 | Gain | MYSM1 TACSTD2 OMA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |