Assay Details
Target Gene Details
Entrez Gene ID: | 22854 |
Gene Name: | netrin G1 |
Gene Aliases: |
Lmnt1 |
Location: |
Chr.1:107139918-107484899 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758957 | Chr.1:106941006 - 107298261 on Build GRCh38 | Loss | PRMT6 NTNG1 |
nsv830982 | Chr.1:107108954 - 107292352 on Build GRCh38 | Gain | NTNG1 |
nsv547330 | Chr.1:107006123 - 107375697 on Build GRCh38 | Gain | PRMT6 NTNG1 |
esv3587104 | Chr.1:107154574 - 107174539 on Build GRCh38 | Gain | NTNG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |