Assay Details
Target Gene Details
Entrez Gene ID: | 54802 |
Gene Name: | tRNA isopentenyltransferase 1 |
Gene Aliases: |
GRO1, IPPT, IPT, IPTase, MOD5, hGRO1 |
Location: |
Chr.1:39841031-39883511 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss | LOC101929536 PPT1 MFSD2A OXCT2 BMP8B RLF MYCL TRIT1 PPIE CAP1 |
esv2666580 | Chr.1:39809257 - 39898016 on Build GRCh38 | Deletion | MYCL TRIT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |