Assay Details
Target Gene Details
Entrez Gene ID: | 92002 |
Gene Name: | family with sequence similarity 58 member A |
Gene Aliases: |
STAR |
Location: |
Chr.X:153587925-153599177 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM58A | NM_001130997.2 | NP_001124469.1 | ||
NM_152274.4 | NP_689487.2 | |||
XM_005277920.4 | XP_005277977.1 | |||
XM_005277921.4 | XP_005277978.1 | |||
XM_011531214.2 | XP_011529516.1 | |||
XM_011531215.2 | XP_011529517.1 | |||
BC001909.1 | ||||
BC007232.1 | ||||
BC032121.2 | AAH32121.1 | |||
BC071851.1 | AAH71851.1 | |||
DQ323993.1 | ABC88595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | MIR3202-1 LOC105373383 HCFC1-AS1 MECP2 IRAK1 SRPK3 MIR6858 LOC105373386 LOC107987332 TMEM187 BGN L1CAM ZFP92 MAGEA1 FLNA PDZD4 LOC105373377 MIR3202-2 HCFC1 ATP6AP1 CH17-340M24.3 OPN1LW OPN1MW PLXNB3 SLC10A3 TEX28 PNMA6A TREX2 ATP2B3 AVPR2 OPN1MW3 LOC107983954 ARHGAP4 SLC6A8 ABCD1 LOC105373378 GDI1 LOC105373381 FAM3A FAM58A MIR718 DNASE1L1 LCA10 FAM50A DUSP9 PNCK CTAG1A RPL10 SNORA70 TKTL1 SSR4 IDH3G TAZ EMD PLXNA3 LAGE3 G6PD BCAP31 HAUS7 OPN1MW2 ZNF275 UBL4A RENBP IKBKG FAM223A NAA10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |