Assay Details
Target Gene Details
Entrez Gene ID: | 8226 |
Gene Name: | pseudouridine 5'-phosphatase |
Gene Aliases: |
DXF68S1E, FAM16AX, GS1, HDHD1, HDHD1A |
Location: |
Chr.X:6768840-7148190 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PUDP |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv265e55 | Chr.X:6471402 - 6918342 on Build GRCh38 | Gain | VCX3A PUDP |
esv3576734 | Chr.X:6537111 - 7202526 on Build GRCh38 | Gain | STS PUDP MIR4767 |
dgv73n64 | Chr.X:6540125 - 6939189 on Build GRCh38 | Gain | PUDP |