Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | SNORA56 CLIC2 FAM223A IKBKG MIR6858 GDI1 F8 MIR718 DNASE1L1 TAZ LOC101927830 LAGE3 SNORA36A ATP6AP1 MIR1184-1 SNORA70 LOC107987332 OPN1MW2 SMIM9 TMLHE CTAG1B TMEM187 F8A2 BRCC3 FAM3A RPL10 H2AFB3 RAB39B H2AFB1 FAM223B SLC10A3 HCFC1 HCFC1-AS1 OPN1MW3 OPN1LW MTCP1 MIR3202-1 GAB3 CTAG1A IRAK1 MECP2 G6PD PLXNA3 F8A3 CTAG2 LOC105373386 VBP1 SPRY3 MPP1 CH17-340M24.3 OPN1MW TKTL1 EMD DKC1 TMLHE-AS1 UBL4A H2AFB2 MIR3202-2 CMC4 FUNDC2 MIR664B FAM50A TEX28 MIR1184-2 F8A1 FLNA MIR1184-3 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | TMLHE SPRY3 |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | TMLHE SPRY3 |
nsv471607 | Chr.X:155652457 - 155812331 on Build GRCh38 | Gain+Loss | SPRY3 |
esv2657377 | Chr.X:155641695 - 155958434 on Build GRCh38 | Deletion | SPRY3 |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | SMIM9 TMLHE TMLHE-AS1 CLIC2 MTCP1 F8A2 BRCC3 H2AFB2 CMC4 FUNDC2 F8 F8A3 H2AFB3 RAB39B LOC101927830 MIR1184-2 H2AFB1 F8A1 VBP1 SPRY3 MPP1 MIR1184-3 MIR1184-1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |