Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | UBL4A MIR3202-2 SMIM9 HCFC1 EMD CH17-340M24.3 FAM223A F8A3 TMEM187 MIR3202-1 CTAG1A CTAG1B OPN1MW DKC1 OPN1MW2 IRAK1 GDI1 FAM3A TAZ RAB39B H2AFB1 SNORA56 MIR664B MECP2 LOC105373386 LOC101927830 H2AFB3 TMLHE F8A1 MTCP1 F8 TKTL1 OPN1LW BRCC3 VBP1 LOC107987332 HCFC1-AS1 SNORA36A DNASE1L1 ATP6AP1 G6PD F8A2 IKBKG MIR6858 MIR1184-2 CLIC2 SLC10A3 FLNA OPN1MW3 CTAG2 FAM223B H2AFB2 LAGE3 MIR1184-1 GAB3 FAM50A TEX28 TMLHE-AS1 RPL10 PLXNA3 MIR718 MIR1184-3 CMC4 FUNDC2 SNORA70 SPRY3 MPP1 |
nsv510870 | Chr.X:155686374 - 155710408 on Build GRCh38 | Deletion | SPRY3 |
esv3575080 | Chr.X:155683281 - 155699244 on Build GRCh38 | Loss | SPRY3 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | SPRY3 TMLHE |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | SPRY3 TMLHE |
nsv471607 | Chr.X:155652457 - 155812331 on Build GRCh38 | Gain+Loss | SPRY3 |
esv2657377 | Chr.X:155641695 - 155958434 on Build GRCh38 | Deletion | SPRY3 |
nsv511663 | Chr.X:155687184 - 155709357 on Build GRCh38 | Gain | SPRY3 |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | H2AFB2 RAB39B H2AFB1 MIR1184-1 TMLHE-AS1 SMIM9 F8A2 LOC101927830 MIR1184-2 F8A3 CLIC2 H2AFB3 TMLHE F8A1 MTCP1 F8 MIR1184-3 CMC4 FUNDC2 BRCC3 VBP1 SPRY3 MPP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |