Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | MPP1 MTCP1 FAM3A CTAG1A OPN1MW F8A1 CLIC2 TMLHE-AS1 EMD PLXNA3 IRAK1 OPN1LW H2AFB2 CH17-340M24.3 LAGE3 VBP1 FLNA ATP6AP1 SNORA56 IKBKG LOC101927830 CTAG1B MIR718 FAM50A LOC105373386 FUNDC2 MECP2 TEX28 TAZ G6PD MIR1184-1 F8 H2AFB1 CMC4 TMLHE LOC107987332 TMEM187 FAM223B MIR6858 MIR3202-1 SMIM9 MIR1184-2 MIR3202-2 SLC10A3 CTAG2 UBL4A H2AFB3 RAB39B GDI1 DNASE1L1 SPRY3 HCFC1 FAM223A SNORA36A DKC1 MIR664B TKTL1 MIR1184-3 HCFC1-AS1 RPL10 OPN1MW3 BRCC3 SNORA70 F8A2 F8A3 OPN1MW2 GAB3 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | TMLHE SPRY3 |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | TMLHE SPRY3 |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | MPP1 SMIM9 MTCP1 LOC101927830 MIR1184-2 MIR1184-3 F8A1 CLIC2 TMLHE-AS1 FUNDC2 H2AFB3 MIR1184-1 RAB39B F8 BRCC3 F8A2 H2AFB1 CMC4 F8A3 TMLHE SPRY3 H2AFB2 VBP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |