Assay Details
Target Gene Details
Entrez Gene ID: | 10587 |
Gene Name: | thioredoxin reductase 2 |
Gene Aliases: |
SELZ, TR, TR-BETA, TR3, TRXR2 |
Location: |
Chr.22:19875518-19941992 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TXNRD2 | NM_006440.4 | NP_006431.2 | ||
AB019694.1 | BAA77601.2 | |||
AB019695.1 | BAA77602.2 | |||
AB209724.1 | BAD92961.1 | |||
AF044212.1 | AAD25167.1 | |||
AF106697.1 | AAD19597.1 | |||
AF166126.1 | AAF21431.1 | |||
AF166127.1 | AAF21432.1 | |||
AF171054.1 | AAD51324.1 | |||
AF201385.1 | AAG47635.1 | |||
AK094698.1 | ||||
AK097708.1 | ||||
BC007489.2 | AAH07489.3 | |||
BC117354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834129 | Chr.22:19705617 - 19919068 on Build GRCh38 | Loss | SEPT5 SEPT5-GP1BB TXNRD2 GNB1L C22orf29 TBX1 GP1BB |
esv3893434 | Chr.22:19470889 - 20343372 on Build GRCh38 | Gain | MIR4761 MIR185 LOC284865 CLDN5 ARVCF LOC440792 COMT DGCR8 SEPT5-GP1BB MIR3618 TANGO2 UFD1L LOC101927859 TRMT2A DGCR6L SEPT5 MIR1286 LINC00896 CCDC188 TXNRD2 CDC45 MIR1306 TBX1 RTN4R MIR6816 RANBP1 ZDHHC8 GNB1L C22orf29 GP1BB LINC00895 |
nsv828938 | Chr.22:19691069 - 20324465 on Build GRCh38 | Gain | MIR4761 MIR185 LOC284865 ARVCF LOC440792 COMT DGCR8 SEPT5-GP1BB MIR3618 TANGO2 TRMT2A DGCR6L SEPT5 MIR1286 LINC00896 CCDC188 TXNRD2 MIR1306 TBX1 RTN4R MIR6816 RANBP1 ZDHHC8 GNB1L C22orf29 GP1BB |
nsv828939 | Chr.22:19696032 - 20324465 on Build GRCh38 | Loss | MIR4761 MIR185 LOC284865 ARVCF LOC440792 COMT DGCR8 SEPT5-GP1BB MIR3618 TANGO2 TRMT2A DGCR6L SEPT5 MIR1286 LINC00896 CCDC188 TXNRD2 MIR1306 TBX1 RTN4R MIR6816 RANBP1 ZDHHC8 GNB1L C22orf29 GP1BB |
nsv515664 | Chr.22:19873352 - 19892114 on Build GRCh38 | Loss | TXNRD2 |
nsv834130 | Chr.22:19837923 - 19940567 on Build GRCh38 | Loss | TXNRD2 GNB1L C22orf29 |
esv3568260 | Chr.22:19777375 - 19881883 on Build GRCh38 | Loss | TXNRD2 GNB1L C22orf29 TBX1 |
esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain | MIR4761 DGCR14 CLDN5 PI4KAP1 DGCR5 MRPL40 CLTCL1 COMT USP18 LOC100996401 SEPT5-GP1BB LOC642643 LINC01311 LOC100996432 DGCR2 GSC2 UFD1L GGT3P FAM230A SEPT5 SLC25A1 DGCR9 TXNRD2 RIMBP3 CDC45 DGCR6 TBX1 LOC102725072 PRODH TMEM191B HIRA C22orf39 GNB1L DGCR11 C22orf29 DGCR10 TSSK2 LOC100996415 GP1BB LOC729461 LINC00895 |
nsv953025 | Chr.22:19869078 - 19937977 on Build GRCh38 | Deletion | TXNRD2 |
dgv7983n54 | Chr.22:19762002 - 20321870 on Build GRCh38 | Gain | MIR4761 MIR185 TRMT2A DGCR6L LOC284865 MIR1286 LINC00896 ARVCF LOC440792 CCDC188 TXNRD2 COMT MIR1306 TBX1 DGCR8 RTN4R MIR6816 RANBP1 MIR3618 ZDHHC8 GNB1L C22orf29 TANGO2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |