Assay Details
Target Gene Details
Entrez Gene ID: | 4689 |
Gene Name: | neutrophil cytosolic factor 4 |
Gene Aliases: |
CGD3, NCF, P40PHOX, SH3PXD4 |
Location: |
Chr.22:36860988-36878017 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCF4 | NM_000631.4 | NP_000622.2 | ||
NM_013416.3 | NP_038202.2 | |||
XM_017028808.1 | XP_016884297.1 | |||
AB025219.1 | BAA89791.1 | |||
AB025220.1 | BAA89792.1 | |||
AK223324.1 | BAD97044.1 | |||
AK290924.1 | ||||
BC002798.1 | AAH02798.1 | |||
BT007346.1 | AAP36010.1 | |||
BX355636.2 | ||||
CR456528.1 | CAG30414.1 | |||
CR542078.1 | CAG46875.1 | |||
X77094.1 | CAA54372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv459881 | Chr.22:36862461 - 36875760 on Build GRCh38 | Loss | NCF4 |
nsv3619 | Chr.22:36856525 - 36877665 on Build GRCh38 | Insertion | NCF4 |
esv2758837 | Chr.22:36779110 - 36964864 on Build GRCh38 | Gain+Loss | NCF4 PVALB CSF2RB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |