Assay Details
Target Gene Details
Entrez Gene ID: | 55615 |
Gene Name: | proline rich 5 |
Gene Aliases: |
FLJ20185k, PP610, PROTOR-1, PROTOR1 |
Location: |
Chr.22:44668547-44737681 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRR5 | NM_001017528.2 | NP_001017528.1 | ||
NM_001017529.2 | NP_001017529.1 | |||
NM_001017530.1 | NP_001017530.1 | |||
NM_001198721.1 | NP_001185650.1 | |||
NM_015366.3 | NP_056181.2 | |||
NM_181333.3 | NP_851850.1 | |||
AF177331.1 | AAG17975.1 | |||
AK055848.1 | ||||
AK091884.1 | ||||
AK291564.1 | ||||
BI825567.1 |
Target Gene Details
Entrez Gene ID: | 553158 |
Gene Name: | PRR5-ARHGAP8 readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:44702198-44862784 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRR5-ARHGAP8 | NM_181334.5 | NP_851851.3 | ||
AL355192.1 | CAB90248.1 | |||
BI488517.1 | ||||
BX335168.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv817999 | Chr.22:44698163 - 44737460 on Build GRCh38 | Gain | PRR5-ARHGAP8 PRR5 |
nsv829268 | Chr.22:44640618 - 44785937 on Build GRCh38 | Gain | PRR5-ARHGAP8 PRR5 ARHGAP8 |
nsv9917 | Chr.22:44706506 - 44707994 on Build GRCh38 | Loss | PRR5-ARHGAP8 PRR5 |
esv2758842 | Chr.22:44630924 - 44814649 on Build GRCh38 | Loss | PRR5-ARHGAP8 PRR5 ARHGAP8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |