Assay Details
Target Gene Details
Entrez Gene ID: | 84271 |
Gene Name: | DNA polymerase delta interacting protein 3 |
Gene Aliases: |
PDIP46, SKAR |
Location: |
Chr.22:42583721-42614962 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLDIP3 | NM_001278657.1 | NP_001265586.1 | ||
NM_032311.4 | NP_115687.2 | |||
NM_178136.2 | NP_835237.1 | |||
NR_103820.1 | ||||
XM_011530457.1 | XP_011528759.1 | |||
XM_017029010.1 | XP_016884499.1 | |||
AB055760.2 | ||||
AK024626.1 | ||||
AK291623.1 | ||||
AK291774.1 | ||||
AK297532.1 | ||||
AK299751.1 | ||||
AK303419.1 | ||||
AL137743.1 | CAH56378.1 | |||
AL160111.1 | CAB77058.1 | |||
AL160112.1 | CAB77059.1 | |||
AY506562.1 | AAR91612.1 | |||
BC001488.2 | AAH01488.2 | |||
BC019643.1 | AAH19643.1 | |||
BC049840.1 | AAH49840.1 | |||
BC056912.1 | AAH56912.2 | |||
BC095411.1 | AAH95411.1 | |||
CR456456.1 | CAG30342.1 | |||
DQ887818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524028 | Chr.22:42592111 - 42631818 on Build GRCh38 | Loss | POLDIP3 RNU12 CYB5R3 |
esv2758839 | Chr.22:42371195 - 42687300 on Build GRCh38 | Gain+Loss | RRP7A NFAM1 POLDIP3 ATP5L2 RNU12 RRP7BP SERHL CYB5R3 SERHL2 |
esv28197 | Chr.22:42483195 - 42609208 on Build GRCh38 | Gain | RRP7A POLDIP3 RRP7BP SERHL SERHL2 |
nsv428391 | Chr.22:42371295 - 42698202 on Build GRCh38 | Gain | A4GALT RRP7A NFAM1 POLDIP3 ATP5L2 RNU12 RRP7BP SERHL CYB5R3 SERHL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |