Assay Details
Target Gene Details
Entrez Gene ID: | 7332 |
Gene Name: | ubiquitin conjugating enzyme E2 L3 |
Gene Aliases: |
E2-F1, L-UBC, UBCH7, UbcM4 |
Location: |
Chr.22:21549447-21624034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2L3 | NM_001256355.1 | NP_001243284.1 | ||
NM_001256356.1 | NP_001243285.1 | |||
NM_003347.3 | NP_003338.1 | |||
NR_028436.2 | ||||
NR_046082.1 | ||||
XM_017028935.1 | XP_016884424.1 | |||
AJ000519.1 | CAA04156.1 | |||
AK125396.1 | ||||
AK291083.1 | ||||
AK293179.1 | ||||
AK299985.1 | ||||
AK311761.1 | ||||
BC053368.1 | AAH53368.1 | |||
BU567707.1 | ||||
BU584304.1 | ||||
CD690076.1 | ||||
CR456606.1 | CAG30492.1 | |||
DA271653.1 | ||||
S81003.1 | AAB36017.1 | |||
X92962.1 | CAA63538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428386 | Chr.22:21328921 - 21595800 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3647318 | Chr.22:21567924 - 21611833 on Build GRCh38 | Gain |
![]() |
nsv834145 | Chr.22:21420780 - 21595800 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
dgv4491n100 | Chr.22:21454896 - 21597273 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1160730 | Chr.22:21373849 - 21633730 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4488n100 | Chr.22:21212930 - 21597273 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)