Assay Details
Target Gene Details
Entrez Gene ID: | 51386 |
Gene Name: | eukaryotic translation initiation factor 3 subunit L |
Gene Aliases: |
EIF3EIP, EIF3S11, EIF3S6IP, HSPC021, HSPC025, MSTP005 |
Location: |
Chr.22:37848774-37888782 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF3L | NM_001242923.1 | NP_001229852.1 | ||
NM_016091.3 | NP_057175.1 | |||
XM_005261625.1 | XP_005261682.1 | |||
XM_006724260.3 | XP_006724323.1 | |||
XM_017028814.1 | XP_016884303.1 | |||
AF077207.1 | AAD27002.1 | |||
AF083243.1 | AAD39841.1 | |||
AF109359.1 | AAQ13507.1 | |||
AK027429.1 | ||||
AK056129.1 | ||||
AK093540.1 | ||||
AK098457.1 | BAC05310.1 | |||
AK129648.1 | ||||
AK222496.1 | BAD96216.1 | |||
AK222529.1 | BAD96249.1 | |||
AK293629.1 | ||||
AK298776.1 | ||||
AK301323.1 | ||||
AK302346.1 | ||||
AK315533.1 | ||||
AL357455.1 | ||||
BC001101.2 | AAH01101.1 | |||
BC007510.2 | AAH07510.1 | |||
BC029265.1 | AAH29265.1 | |||
CR456436.1 | CAG30322.1 | |||
DA051005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss | EIF3L MIR659 GALR3 PDXP GGA1 LOC102724378 LGALS1 LOC101927051 H1F0 MIR658 SH3BP1 GCAT ANKRD54 NOL12 TRIOBP |
nsv191391 | Chr.22:37859396 - 37863103 on Build GRCh38 | Deletion | EIF3L |
esv3893482 | Chr.22:37832559 - 37927500 on Build GRCh38 | Gain | EIF3L MIR658 MIR659 ANKRD54 MICALL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |