Assay Details
Target Gene Details
Entrez Gene ID: | 57167 |
Gene Name: | spalt like transcription factor 4 |
Gene Aliases: |
DRRS, HSAL4, ZNF797 |
Location: |
Chr.20:51782717-51802523 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SALL4 | NM_001318031.1 | NP_001304960.1 | ||
NM_020436.4 | NP_065169.1 | |||
XM_011528921.2 | XP_011527223.1 | |||
AK001666.1 | ||||
AY170621.1 | AAO16566.1 | |||
AY170622.1 | AAO16567.1 | |||
AY172738.1 | AAO44950.1 | |||
BC111714.1 | AAI11715.1 | |||
DA666635.1 | ||||
DB066881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758800 | Chr.20:51706078 - 51802856 on Build GRCh38 | Loss | ATP9A SALL4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |