Assay Details
Target Gene Details
Entrez Gene ID: | 149775 |
Gene Name: | GNAS antisense RNA 1 |
Gene Aliases: |
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG |
Location: |
Chr.20:58818918-58850903 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNAS-AS1 | NR_002785.2 | |||
AJ251759.1 | ||||
BF589977.1 | ||||
DB067618.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)