Assay Details
Target Gene Details
Entrez Gene ID: | 9584 |
Gene Name: | RNA binding motif protein 39 |
Gene Aliases: |
CAPER, CAPERalpha, FSAP59, HCC1, RNPC2 |
Location: |
Chr.20:35703609-35742336 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526188 | Chr.20:35576541 - 36019079 on Build GRCh38 | Gain | ROMO1 CPNE1 PHF20 SCAND1 CNBD2 SPAG4 RBM12 RBM39 NFS1 FER1L4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |