Assay Details
Target Gene Details
Entrez Gene ID: | 4773 |
Gene Name: | nuclear factor of activated T-cells 2 |
Gene Aliases: |
NFAT1, NFATP |
Location: |
Chr.20:51386957-51562857 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFATC2 | NM_001136021.2 | NP_001129493.1 | ||
NM_001258292.1 | NP_001245221.1 | |||
NM_001258294.1 | NP_001245223.1 | |||
NM_001258295.1 | NP_001245224.1 | |||
NM_001258296.1 | NP_001245225.1 | |||
NM_001258297.1 | NP_001245226.1 | |||
NM_012340.4 | NP_036472.2 | |||
NM_173091.3 | NP_775114.1 | |||
XM_017027851.1 | XP_016883340.1 | |||
AK226146.1 | ||||
AK304147.1 | ||||
BC136418.1 | ||||
BC144074.1 | ||||
BF914609.1 | ||||
BU429325.1 | ||||
CF136470.1 | ||||
EU887573.1 | ||||
EU887574.1 | ||||
EU887575.1 | ||||
EU887576.1 | ||||
EU887577.1 | ||||
EU887578.1 | ||||
EU887579.1 | ||||
EU887580.1 | ||||
U43342.1 | AAC50887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2751917 | Chr.20:51052156 - 51598401 on Build GRCh38 | Gain | ATP9A MIR3194 NFATC2 |
nsv1055542 | Chr.20:51378130 - 51402077 on Build GRCh38 | Gain | NFATC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |