Assay Details
Target Gene Details
Entrez Gene ID: | 57333 |
Gene Name: | reticulocalbin 3 |
Gene Aliases: |
RLP49 |
Location: |
Chr.19:49527579-49543638 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RCN3 | NM_020650.2 | NP_065701.2 | ||
XM_005259089.3 | XP_005259146.1 | |||
XM_011527143.1 | XP_011525445.1 | |||
XM_017027023.1 | XP_016882512.1 | |||
AF183423.1 | AAG09692.1 | |||
AY195859.1 | AAO43054.1 | |||
AY358423.1 | AAQ88789.1 | |||
BC013436.1 | AAH13436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | C19orf68 RCN3 SNORD35B GRIN2D LHB BCL2L12 SNAR-A10 FLT3LG CGB2 PTOV1-AS1 SCAF1 HSD17B14 NUP62 ADM5 SLC6A16 RRAS CA11 ELSPBP1 PTH2 SNAR-A6 KCNJ14 SNAR-C2 LIG1 MIR6800 SPACA4 TEAD2 PRRG2 BCAT2 SIGLEC11 FLJ26850 SNORD34 LOC105447645 FTL LOC101059948 MIR6798 SNAR-B1 CPT1C SULT2B1 RPL18 TRPM4 PLA2G4C-AS1 ALDH16A1 CARD8 SNAR-B2 FUZ SLC17A7 PNKP KDELR1 SNAR-A14 DHDH PLA2G4C LOC101928295 RPL13A SNAR-C3 ZNF114 MIR4749 SNAR-G2 SNAR-A13 PTOV1-AS2 CGB8 MAMSTR LOC105372430 VRK3 RASIP1 MIR4750 MIR150 TSKS LOC100287477 AKT1S1 PRMT1 NOSIP BSPH1 NUCB1 SNAR-A12 PLEKHA4 CD37 CARD8-AS1 GYS1 MIR4751 LMTK3 SIGLEC16 PPFIA3 CCDC114 HRC TBC1D17 NTF4 IL4I1 TULP2 SNAR-A1 DKKL1 FUT2 AP2A1 GRWD1 CABP5 PPP1R15A SNAR-A8 SNAR-A7 CGB1 SNAR-C4 CGB5 MIR6799 RUVBL2 TMEM143 DBP NUCB1-AS1 SNAR-A5 SNAR-C1 NTN5 ZNF473 SNAR-A9 CGB3 PRR12 SNRNP70 C19orf73 MED25 SYNGR4 FAM83E FUT1 CCDC155 ATF5 SNAR-A2 SNAR-A4 RPS11 SPHK2 CYTH2 SNAR-D FCGRT IZUMO1 SNAR-A11 PTOV1 GFY SNAR-A3 BAX SNORD32A IRF3 PIH1D1 SNAR-G1 CGB7 SEC1P FGF21 LIN7B SNORD33 SNAR-C5 KCNA7 EMP3 MIR5088 MIR4324 SNORD35A |
nsv833863 | Chr.19:49479177 - 49629716 on Build GRCh38 | Loss | RCN3 SNORD35B FCGRT PRR12 SNORD32A FLT3LG MIR150 PRRG2 SNORD34 NOSIP SNORD33 SNORD35A RPL13A RPS11 |
nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss | RCN3 SNORD35B FCGRT ALDH16A1 SNORD32A PIH1D1 FLT3LG MIR150 SNORD34 NOSIP SNORD33 SNORD35A RPL13A RPS11 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | RCN3 SNORD35B PRR12 BCL2L12 FUZ MED25 PTOV1-AS1 SCAF1 RPL13A RPS11 AP2A1 ADM5 RRAS FCGRT SNORD32A IRF3 MIR6800 MIR150 TSKS PRRG2 MIR6799 SNORD34 PRMT1 NOSIP SNORD33 MIR5088 CPT1C SNORD35A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |