Assay Details
Target Gene Details
Entrez Gene ID: | 10498 |
Gene Name: | coactivator associated arginine methyltransferase 1 |
Gene Aliases: |
PRMT4 |
Location: |
Chr.19:10871577-10923078 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CARM1 | NM_199141.1 | NP_954592.1 | ||
XM_005259708.4 | XP_005259765.1 | |||
AL833242.1 | ||||
BU176535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion | MIR4748 CARM1 SLC44A2 ILF3-AS1 QTRT1 TMED1 C19orf38 DNM2 MIR6793 ILF3 MIR199A1 MIR638 |
nsv833749 | Chr.19:10881255 - 11027578 on Build GRCh38 | Loss | SMARCA4 CARM1 C19orf52 YIPF2 |
nsv833748 | Chr.19:10757002 - 10925368 on Build GRCh38 | Loss | MIR4748 CARM1 YIPF2 TMED1 C19orf38 DNM2 MIR6793 MIR199A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |