Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | NP_005348.2 | ||
XM_005258937.3 | XP_005258994.1 | |||
XM_005258938.4 | XP_005258995.1 | |||
XM_005258939.3 | XP_005258996.2 | |||
XM_005258940.3 | XP_005258997.1 | |||
XM_006723218.2 | XP_006723281.1 | |||
XM_017026810.1 | XP_016882299.1 | |||
AK292482.1 | ||||
BC070041.1 | AAH70041.1 | |||
U40002.1 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 | ||||
DW455857.1 |
Target Gene Details
Entrez Gene ID: | 101930071 |
Gene Name: | uncharacterized LOC101930071 |
Gene Aliases: |
- |
Location: |
Chr.19:42397128-42408452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101930071 | NR_126041.1 | |||
BU566442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | LIPE LIPE-AS1 CNFN LOC101930071 |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | ZNF526 RPS19 MIR8077 LIPE-AS1 PAFAH1B3 LOC101930071 LOC100505622 CIC ARHGEF1 MIR6797 RABAC1 ERF ZNF574 POU2F2 LOC390937 MIR4323 LIPE DEDD2 GRIK5 MEGF8 CD79A TMEM145 PRR19 ATP1A3 GSK3A LOC100505585 CNFN |
nsv579632 | Chr.19:42400501 - 42408251 on Build GRCh38 | Gain | LIPE LIPE-AS1 LOC101930071 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | ZNF526 LIPE DEDD2 MIR8077 LIPE-AS1 MEGF8 CEACAM1 PAFAH1B3 LOC101930071 TMEM145 PRR19 CIC GSK3A CXCL17 ERF POU2F2 CNFN CEACAM8 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | ZNF526 RPS19 MIR8077 LIPE-AS1 PAFAH1B3 LOC101930071 LOC100505622 CIC CEACAM6 ARHGEF1 MIR6797 RABAC1 ERF ZNF574 POU2F2 CEACAM5 LOC390937 MIR4323 LIPE DEDD2 CEACAM3 DMRTC2 GRIK5 MEGF8 CEACAM4 CD79A LYPD4 TMEM145 PRR19 ATP1A3 GSK3A LOC100505585 CEACAM7 CNFN |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | LIPE PSG6 MIR8077 LIPE-AS1 MEGF8 PSG2 PSG3 CEACAM1 PSG10P PSG7 PAFAH1B3 LOC101930071 LOC100289650 TMEM145 PRR19 CIC PSG1 GSK3A CXCL17 PSG11 ERF PSG8 CNFN CEACAM8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |