Assay Details
Target Gene Details
Entrez Gene ID: | 976 |
Gene Name: | adhesion G protein-coupled receptor E5 |
Gene Aliases: |
CD97, TM7LN1 |
Location: |
Chr.19:14381144-14408725 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRE5 | NM_001025160.2 | NP_001020331.1 | ||
NM_001784.4 | NP_001775.2 | |||
NM_078481.3 | NP_510966.1 | |||
XM_011528451.2 | XP_011526753.1 | |||
XM_017027547.1 | XP_016883036.1 | |||
AK097264.1 | ||||
AK225655.1 | ||||
AK225677.1 | ||||
AK292159.1 | ||||
AK300343.1 | ||||
AK304553.1 | ||||
AK308368.1 | ||||
AK314697.1 | ||||
AX775811.1 | CAE11605.1 | |||
AX775813.1 | CAE11606.1 | |||
BC026690.2 | AAH26690.1 | |||
BM545710.1 | ||||
CB127827.1 | ||||
DC381605.1 | ||||
DC414739.1 | ||||
U76764.1 | AAB36682.1 | |||
X84700.1 | CAA59173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833764 | Chr.19:14381787 - 14504061 on Build GRCh38 | Loss | DDX39A GIPC1 SNRPGP15 ADGRE5 PTGER1 PKN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |