Assay Details
Target Gene Details
Entrez Gene ID: | 94030 |
Gene Name: | leucine rich repeat containing 4B |
Gene Aliases: |
HSM, LRIG4 |
Location: |
Chr.19:50516892-50568208 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC4B | NM_001080457.1 | NP_001073926.1 | ||
XM_005259429.4 | XP_005259486.1 | |||
XM_006723506.3 | XP_006723569.1 | |||
XM_011527520.2 | XP_011525822.1 | |||
BC032460.1 | ||||
BC131897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953604 | Chr.19:50506544 - 50580143 on Build GRCh38 | Deletion | ASPDH JOSD2 LRRC4B |
nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion | ASPDH EMC10 MYBPC2 JOSD2 LRRC4B FAM71E1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |