Assay Details
Target Gene Details
Entrez Gene ID: | 64377 |
Gene Name: | carbohydrate sulfotransferase 8 |
Gene Aliases: |
GALNAC4ST1, GalNAc4ST, PSS3 |
Location: |
Chr.19:33621955-33773509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHST8 | NM_001127895.1 | NP_001121367.1 | ||
NM_001127896.1 | NP_001121368.1 | |||
AB047801.1 | BAB19806.1 | |||
BC011380.1 | AAH11380.1 | |||
BC014250.2 | AAH14250.1 | |||
BI917117.1 | ||||
BX354555.2 | ||||
DB500322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv139e55 | Chr.19:33406261 - 34032878 on Build GRCh38 | Gain | PEPD LOC100996681 CHST8 KCTD15 |
esv2758755 | Chr.19:33286168 - 34090656 on Build GRCh38 | Gain | PEPD CEBPG LOC100996681 CEBPA-AS1 CHST8 KCTD15 CEBPA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |