Assay Details
Target Gene Details
Entrez Gene ID: | 5424 |
Gene Name: | DAN polymerase delta 1, catalytic subunit |
Gene Aliases: |
CDC2, CRCS10, MDPL, POLD |
Location: |
Chr.19:50384291-50418018 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLD1 | NM_001256849.1 | NP_001243778.1 | ||
NM_002691.3 | NP_002682.2 | |||
NR_046402.1 | ||||
XM_005259008.4 | XP_005259065.1 | |||
XM_017026882.1 | XP_016882371.1 | |||
AB209560.1 | BAD92797.1 | |||
BC008800.1 | AAH08800.1 | |||
BM193218.1 | ||||
CD101690.1 | ||||
M80397.1 | AAA58439.1 | |||
M81735.1 | AAA35768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066389 | Chr.19:50319151 - 50453251 on Build GRCh38 | Gain | MYBPC2 NAPSB SPIB POLD1 NR1H2 KCNC3 NAPSA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |