Assay Details
Target Gene Details
Entrez Gene ID: | 6929 |
Gene Name: | transcription factor 3 |
Gene Aliases: |
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21 |
Location: |
Chr.19:1609290-1652590 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | PLK5 PCSK4 ADAMTSL5 MUM1 CSNK1G2 KLF16 NDUFS7 TCF3 DAZAP1 RPS15 CSNK1G2-AS1 MEX3D ATP8B3 REEP6 LOC100288123 BTBD2 UQCR11 MIR1909 C19orf25 MBD3 ADAT3 REXO1 ABHD17A APC2 ONECUT3 SCAMP4 GAMT |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | PLK5 PCSK4 ADAMTSL5 MUM1 CSNK1G2 MKNK2 KLF16 NDUFS7 TCF3 DAZAP1 RPS15 CSNK1G2-AS1 MEX3D ATP8B3 REEP6 LOC100288123 BTBD2 UQCR11 MIR1909 C19orf25 MBD3 ADAT3 REXO1 ABHD17A APC2 ONECUT3 SCAMP4 GAMT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |