Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
AK124343.1 | ||||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | LOC105447645 FCGRT SNAR-A9 MIR150 IRF3 MIR4750 NOSIP DHDH LOC105372430 BCAT2 SNAR-A5 GRIN2D EMP3 RPL13A ADM5 DKKL1 FUZ SNAR-B1 FTL LOC100287477 BCL2L12 ELSPBP1 SNORD35B NUP62 SNAR-A1 SNAR-G2 MIR6798 PPFIA3 ALDH16A1 PRRG2 KDELR1 FLT3LG BAX RASIP1 NUCB1-AS1 MIR4749 PIH1D1 SNORD34 PLA2G4C CGB2 FUT2 TEAD2 AKT1S1 SEC1P CA11 LOC101928295 MIR5088 SNORD35A PPP1R15A CGB8 RPS11 VRK3 SPACA4 SCAF1 SNAR-C1 MIR6800 SNAR-C5 SNAR-A4 KCNJ14 PRMT1 NTN5 SIGLEC11 RRAS LHB SULT2B1 SNAR-A7 PLA2G4C-AS1 CABP5 LOC101059948 SNAR-C2 SNORD33 SNAR-A2 NTF4 ATF5 SNAR-A14 SNAR-A10 DBP GYS1 SNAR-D CGB7 MIR4751 PTOV1-AS1 CGB1 SNRNP70 MIR4324 FUT1 GRWD1 SYNGR4 MIR6799 PNKP KCNA7 SNAR-A3 SLC17A7 PTH2 ZNF473 CYTH2 CARD8-AS1 BSPH1 GFY AP2A1 SIGLEC16 CGB5 PRR12 LIG1 CPT1C SNAR-C3 ZNF114 RPL18 TMEM143 CARD8 CD37 FLJ26850 SNAR-A12 TBC1D17 RCN3 SLC6A16 C19orf68 CCDC155 LMTK3 SNAR-B2 IZUMO1 FGF21 MAMSTR RUVBL2 PTOV1 HRC SNAR-G1 SPHK2 C19orf73 SNAR-C4 LIN7B IL4I1 FAM83E SNAR-A13 TRPM4 HSD17B14 SNAR-A6 SNAR-A8 CGB3 TSKS PTOV1-AS2 CCDC114 TULP2 SNAR-A11 PLEKHA4 MED25 SNORD32A NUCB1 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | MIR6799 BCL2L12 PRMT1 MIR5088 IRF3 TSKS PTOV1-AS2 PTOV1-AS1 AP2A1 RRAS PTOV1 PRR12 CPT1C MED25 SCAF1 ADM5 MIR6800 MIR4749 FUZ |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | MIR6799 PNKP AKT1S1 ATF5 IL4I1 TBC1D17 NUP62 MIR4750 TSKS PTOV1-AS2 MIR4751 PTOV1-AS1 AP2A1 PTOV1 MED25 MIR6800 MIR4749 FUZ |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | AKT1S1 ATF5 TBC1D17 MIR5088 IRF3 MIR4750 NOSIP MIR4751 PTOV1-AS1 PTOV1 SCAF1 ADM5 MIR6800 FUZ MIR6799 PNKP BCL2L12 PRMT1 IL4I1 NUP62 TSKS PTOV1-AS2 PRRG2 AP2A1 RRAS PRR12 CPT1C MED25 MIR4749 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | FCGRT MIR5088 RCN3 MIR150 SNORD35A IRF3 NOSIP PTOV1-AS1 RPS11 RPL13A SCAF1 ADM5 MIR6800 FUZ MIR6799 BCL2L12 PRMT1 SNORD35B TSKS PRRG2 AP2A1 RRAS PRR12 CPT1C MED25 SNORD32A SNORD34 SNORD33 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |