Assay Details
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 | ||||
DW455857.1 |
Target Gene Details
Entrez Gene ID: | 101930071 |
Gene Name: | uncharacterized LOC101930071 |
Gene Aliases: |
- |
Location: |
Chr.19:42397128-42408452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101930071 | NR_126041.1 | |||
BU566442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | CNFN LIPE-AS1 LOC101930071 LIPE |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | RABAC1 CD79A DEDD2 POU2F2 ZNF526 ATP1A3 LOC100505622 ARHGEF1 GSK3A LIPE-AS1 LOC101930071 PRR19 MIR8077 LOC390937 CNFN MEGF8 TMEM145 ERF MIR6797 MIR4323 GRIK5 LOC100505585 RPS19 ZNF574 PAFAH1B3 CIC LIPE |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | PRR19 MIR8077 CNFN MEGF8 CEACAM1 DEDD2 POU2F2 TMEM145 ERF ZNF526 CXCL17 GSK3A LIPE-AS1 CEACAM8 PAFAH1B3 CIC LOC101930071 LIPE |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | CEACAM6 RABAC1 CD79A DEDD2 POU2F2 ZNF526 ATP1A3 LOC100505622 ARHGEF1 GSK3A LIPE-AS1 CEACAM4 CEACAM7 LOC101930071 DMRTC2 PRR19 MIR8077 LOC390937 CNFN MEGF8 TMEM145 ERF MIR6797 MIR4323 GRIK5 CEACAM5 LOC100505585 CEACAM3 RPS19 ZNF574 PAFAH1B3 CIC LYPD4 LIPE |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | PRR19 MIR8077 CNFN PSG7 PSG1 MEGF8 CEACAM1 TMEM145 ERF PSG8 PSG2 PSG10P PSG3 CXCL17 LOC100289650 PSG6 GSK3A LIPE-AS1 CEACAM8 PSG11 PAFAH1B3 CIC LOC101930071 LIPE |