Assay Details
Target Gene Details
Entrez Gene ID: | 80816 |
Gene Name: | additional sex combs like 3, transcriptional regulator |
Gene Aliases: |
BRPS, KIAA1713 |
Location: |
Chr.18:33578577-33751195 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASXL3 | NM_030632.2 | NP_085135.1 | ||
XM_005258356.1 | XP_005258413.1 | |||
XM_011526209.1 | XP_011524511.1 | |||
AK131454.1 | BAD18599.1 | |||
BE145544.1 | ||||
DB061724.1 | ||||
DB090653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762001 | Chr.18:33537508 - 33709933 on Build GRCh38 | Gain | ASXL3 |
esv2668525 | Chr.18:33248704 - 33887857 on Build GRCh38 | Deletion | ASXL3 NOL4 CCDC178 |
nsv833618 | Chr.18:33417973 - 33593061 on Build GRCh38 | Gain | ASXL3 CCDC178 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |