Assay Details
Target Gene Details
Entrez Gene ID: | 5338 |
Gene Name: | phospholipase D2 |
Gene Aliases: |
- |
Location: |
Chr.17:4807101-4823432 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLD2 | NM_001243108.1 | NP_001230037.1 | ||
NM_002663.4 | NP_002654.3 | |||
XM_005256696.2 | XP_005256753.1 | |||
XM_017024764.1 | XP_016880253.1 | |||
AB209374.1 | BAD92611.1 | |||
AF033850.1 | AAD04197.1 | |||
AF035483.1 | AAC24498.1 | |||
AF038440.1 | AAB96655.1 | |||
AF038441.1 | AAB96656.1 | |||
AK304246.1 | ||||
AK314797.1 | ||||
AY178289.1 | AAO21120.1 | |||
BC015033.1 | AAH15033.1 | |||
BC056871.1 | AAH56871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833347 | Chr.17:4785297 - 4955981 on Build GRCh38 | Loss | RNF167 GLTPD2 MINK1 SLC25A11 PFN1 PLD2 VMO1 ENO3 GP1BA PSMB6 C17orf107 CHRNE LOC107984973 |
esv3639795 | Chr.17:4812578 - 4820832 on Build GRCh38 | Loss | PLD2 |
nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain | ALOX15 LOC101928000 DHX33 MED11 RNF167 MIR6865 MINK1 SLC25A11 PFN1 ZNF594 LOC101927979 RPAIN KIF1C PLD2 NUP88 ENO3 SMTNL2 GP1BA PSMB6 CXCL16 C17orf107 LOC107984973 ZFP3 RABEP1 ZMYND15 LOC100130950 SCIMP INCA1 TM4SF5 C1QBP GLTPD2 SLC52A1 ZNF232 PELP1 VMO1 LOC101559451 LOC400568 USP6 ARRB2 MIR6864 CAMTA2 SPAG7 CHRNE LOC102724009 |
nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss | ZMYND15 MED11 TM4SF5 GLTPD2 MINK1 PLD2 PELP1 VMO1 LOC101559451 ARRB2 PSMB6 CXCL16 LOC107984973 |
nsv820043 | Chr.17:4810769 - 4814353 on Build GRCh38 | Loss | PLD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |