Assay Details
Target Gene Details
Entrez Gene ID: | 6362 |
Gene Name: | C-C motif chemokine ligand 18 |
Gene Aliases: |
AMAC-1, AMAC1, CKb7, DC-CK1, DCCK1, MIP-4, PARC, SCYA18 |
Location: |
Chr.17:36064272-36071481 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL18 | NM_002988.3 | NP_002979.1 | ||
AB000221.1 | BAA21670.1 | |||
AB451458.1 | ||||
BC069700.1 | AAH69700.1 | |||
BC096124.3 | AAH96124.1 | |||
BC096125.3 | AAH96125.1 | |||
BC096126.1 | AAH96126.1 | |||
BC096127.1 | AAH96127.1 | |||
CR407660.1 | CAG28588.1 | |||
Y13710.1 | CAA74039.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762388 | Chr.17:36060272 - 36265379 on Build GRCh38 | Gain+Loss | CCL3 CCL18 CCL3L3 CCL4 TBC1D3I TBC1D3B CCL4L2 |
nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain | CCL3 CCL18 CCL4 CCL15-CCL14 HEATR9 RDM1 LRRC37A8P LOC101927339 CCL16 CCL15 CCL14 LYZL6 CCL23 TAF15 CCL5 |
nsv574755 | Chr.17:36025629 - 36153146 on Build GRCh38 | Gain | CCL3 CCL18 CCL4 |
nsv833427 | Chr.17:35958281 - 36115227 on Build GRCh38 | Loss | CCL3 LOC101927339 CCL18 CCL16 CCL15 CCL14 CCL4 CCL15-CCL14 CCL23 |
dgv3143n100 | Chr.17:36060260 - 36134107 on Build GRCh38 | Gain | CCL3 CCL18 CCL4 |
dgv120e55 | Chr.17:36044950 - 36150005 on Build GRCh38 | Gain | CCL3 CCL18 CCL4 |
nsv833424 | Chr.17:35911883 - 36092128 on Build GRCh38 | Gain | CCL3 LOC101927339 CCL18 CCL16 CCL15 CCL14 LYZL6 CCL15-CCL14 CCL23 RDM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |