Assay Details
Target Gene Details
Entrez Gene ID: | 201140 |
Gene Name: | dehydrogenase/reductase 7C |
Gene Aliases: |
SDR32C2 |
Location: |
Chr.17:9771434-9791297 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHRS7C | NM_001105571.2 | NP_001099041.1 | ||
NM_001220493.1 | NP_001207422.1 | |||
BC147024.1 | ||||
BC147025.1 | ||||
BC171909.1 | ||||
DN831946.1 | ||||
DN831947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833360 | Chr.17:9734634 - 9900986 on Build GRCh38 | Loss | GLP2R GSG1L2 RCVRN DHRS7C |
nsv833358 | Chr.17:9643743 - 9828246 on Build GRCh38 | Gain | GLP2R GSG1L2 USP43 DHRS7C |
esv3639923 | Chr.17:9778120 - 9786051 on Build GRCh38 | Loss | DHRS7C |
nsv510697 | Chr.17:9729641 - 9812090 on Build GRCh38 | Deletion | GSG1L2 USP43 DHRS7C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |