Assay Details
Target Gene Details
Entrez Gene ID: | 6605 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1 |
Gene Aliases: |
BAF57, CSS5 |
Location: |
Chr.17:40627720-40647851 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMARCE1 | NM_003079.4 | NP_003070.3 | ||
AK001532.1 | ||||
AK095047.1 | ||||
AK294218.1 | ||||
AK294666.1 | ||||
BC007082.1 | AAH07082.1 | |||
BC011017.1 | AAH11017.1 | |||
BC063700.1 | AAH63700.1 | |||
BT007176.1 | AAP35840.1 | |||
EU327017.1 | ||||
EU327018.1 | ||||
EU327019.1 | ||||
EU327020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574994 | Chr.17:40563547 - 40668384 on Build GRCh38 | Gain | KRT222 CCR7 SMARCE1 |
esv2758689 | Chr.17:40592048 - 40693852 on Build GRCh38 | Gain | KRT222 LOC105371775 SMARCE1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |