Assay Details
Target Gene Details
Entrez Gene ID: | 2885 |
Gene Name: | growth factor receptor bound protein 2 |
Gene Aliases: |
ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084, NCKAP2 |
Location: |
Chr.17:75318076-75405709 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRB2 | NM_002086.4 | NP_002077.1 | ||
NM_203506.2 | NP_987102.1 | |||
AF498925.1 | AAM21073.1 | |||
AK091010.1 | ||||
AK315565.1 | ||||
BC000631.2 | AAH00631.1 | |||
BI459414.1 | ||||
CR450363.1 | CAG29359.1 | |||
CR541942.1 | CAG46740.1 | |||
DA458680.1 | ||||
L29511.1 | AAC37549.1 | |||
M96995.1 | AAA58448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv833541 | Chr.17:75297204 - 75460023 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)