Assay Details
Target Gene Details
Entrez Gene ID: | 11269 |
Gene Name: | DEAD-box helicase 19B |
Gene Aliases: |
DBP5, DDX19, RNAh |
Location: |
Chr.16:70289767-70335213 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833271 | Chr.16:70151791 - 70319431 on Build GRCh38 | Gain+Loss | PDPR LOC105371328 LOC100506083 AARS SMG1P7 DDX19B CLEC18C EXOSC6 LOC400541 LOC107984138 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |