Assay Details
Target Gene Details
Entrez Gene ID: | 3029 |
Gene Name: | hydroxyacylglutathione hydrolase |
Gene Aliases: |
GLO2, GLX2, GLXII, HAGH1 |
Location: |
Chr.16:1809099-1827194 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAGH | NM_001040427.1 | NP_001035517.1 | ||
NM_001286249.1 | NP_001273178.1 | |||
NM_005326.4 | NP_005317.2 | |||
XM_011522469.2 | XP_011520771.1 | |||
XM_011522470.2 | XP_011520772.1 | |||
XM_017023195.1 | XP_016878684.1 | |||
AK290155.1 | ||||
AK298174.1 | ||||
AK299173.1 | ||||
AK299993.1 | ||||
AK308825.1 | 5 | 953 | ||
BC000840.1 | AAH00840.1 | |||
BC002627.2 | ||||
BU176541.1 | ||||
X90999.1 | CAA62483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv952900 | Chr.16:1724500 - 1827999 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)