Assay Details
Target Gene Details
Entrez Gene ID: | 25839 |
Gene Name: | component of oligomeric golgi complex 4 |
Gene Aliases: |
CDG2J, COD1 |
Location: |
Chr.16:70480569-70523554 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COG4 | NM_001195139.1 | NP_001182068.1 | ||
NM_015386.2 | NP_056201.2 | |||
XM_011522981.2 | XP_011521283.1 | |||
AB088369.1 | BAC05682.1 | |||
AK022874.1 | BAB14286.1 | |||
AK026435.1 | BAB15483.1 | |||
AK096557.1 | BAC04816.1 | |||
AK293235.1 | ||||
AK297557.1 | ||||
AL050101.1 | CAB43272.1 | |||
AU125729.1 | ||||
BC006306.2 | AAH06306.2 | |||
BC013347.2 | AAH13347.2 | |||
BC027726.2 | AAH27726.1 | |||
BC072438.1 | AAH72438.1 | |||
BC128144.1 | ||||
BC128145.1 | ||||
BX648010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain | VAC14-AS1 FUK VAC14 SNORD111 ST3GAL2 SF3B3 COG4 MTSS1L IL34 SNORD111B |
nsv833272 | Chr.16:70335866 - 70535820 on Build GRCh38 | Loss | DDX19A FUK ST3GAL2 SF3B3 COG4 LOC100506083 SNORD111B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |