Assay Details
Target Gene Details
Entrez Gene ID: | 79834 |
Gene Name: | pseudopodium enriched atypical kinase 1 |
Gene Aliases: |
SGK269 |
Location: |
Chr.15:77100829-77420104 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PEAK1 | NM_024776.3 | NP_079052.2 | ||
XM_005254670.2 | XP_005254727.1 | |||
XM_005254671.2 | XP_005254728.1 | |||
XM_005254673.2 | XP_005254730.1 | |||
XM_005254674.2 | XP_005254731.1 | |||
XM_005254675.4 | XP_005254732.1 | |||
XM_011522033.2 | XP_011520335.1 | |||
XM_011522034.2 | XP_011520336.1 | |||
XM_011522035.2 | XP_011520337.1 | |||
XM_011522036.2 | XP_011520338.1 | |||
XM_011522037.2 | XP_011520339.1 | |||
XM_011522038.2 | XP_011520340.1 | |||
XM_011522039.2 | XP_011520341.1 | |||
XM_011522040.1 | XP_011520342.1 | |||
XM_011522042.2 | XP_011520344.1 | |||
AB082533.1 | BAC02711.1 | |||
AK127658.1 | BAC87076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)