Assay Details
Target Gene Details
Entrez Gene ID: | 100137047 |
Gene Name: | jumonji domain containing 7 |
Gene Aliases: |
- |
Location: |
Chr.15:41828085-41837587 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JMJD7 | NM_001114632.1 | NP_001108104.1 | ||
AK000550.1 | ||||
AK000663.1 | ||||
AK000814.1 | BAA91385.1 | |||
AK225503.1 | ||||
AK289833.1 | ||||
AK290051.1 | ||||
BC025290.1 | AAH25290.1 | |||
BC139899.1 | ||||
DA100207.1 |
Target Gene Details
Entrez Gene ID: | 8681 |
Gene Name: | JMJD7-PLA2G4B readthrough |
Gene Aliases: |
HsT16992, cPLA2-beta |
Location: |
Chr.15:41828085-41848148 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JMJD7-PLA2G4B | NM_001198588.1 | NP_001185517.1 | ||
NM_005090.3 | NP_005081.1 | |||
AB209150.1 | BAD92387.1 | |||
AF065215.1 | AAC78836.1 | |||
AF065216.1 | ||||
AF121908.1 | AAD32135.1 | |||
DA096882.1 | ||||
DA121536.1 | ||||
DQ523799.1 | ||||
DQ523800.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9244 | Chr.15:41818514 - 41916607 on Build GRCh38 | Gain | PLA2G4B JMJD7-PLA2G4B JMJD7 EHD4 MIR4310 SPTBN5 MAPKBP1 LOC105370792 |
nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain | JMJD7-PLA2G4B PLA2G4E PLA2G4D PLA2G4E-AS1 MAPKBP1 LOC105370792 PLA2G4F PLA2G4B JMJD7 EHD4 MIR4310 SPTBN5 EHD4-AS1 |
nsv1506 | Chr.15:41824733 - 41857173 on Build GRCh38 | Insertion | PLA2G4B JMJD7-PLA2G4B JMJD7 SPTBN5 MAPKBP1 |
nsv952613 | Chr.15:41816803 - 41884902 on Build GRCh38 | Deletion | PLA2G4B JMJD7-PLA2G4B JMJD7 MIR4310 SPTBN5 MAPKBP1 |
nsv517631 | Chr.15:41804728 - 41895294 on Build GRCh38 | Loss | PLA2G4B JMJD7-PLA2G4B JMJD7 MIR4310 SPTBN5 MAPKBP1 LOC105370792 |
dgv2582n100 | Chr.15:41791376 - 41900562 on Build GRCh38 | Gain | PLA2G4B JMJD7-PLA2G4B JMJD7 EHD4 MIR4310 SPTBN5 MAPKBP1 LOC105370792 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |