Assay Details
Target Gene Details
Entrez Gene ID: | 10099 |
Gene Name: | tetraspanin 3 |
Gene Aliases: |
TM4-A, TM4SF8, TSPAN-3 |
Location: |
Chr.15:77044017-77071228 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSPAN3 | NM_001168412.1 | NP_001161884.1 | ||
NM_005724.5 | NP_005715.1 | |||
NM_198902.2 | NP_944492.1 | |||
AF054840.1 | AAC69716.1 | |||
AF133423.1 | AAF08362.1 | |||
AK001305.1 | BAA91613.1 | |||
AK001310.1 | BAA91615.1 | |||
AK001326.1 | BAA91627.1 | |||
AK075336.1 | ||||
AK075482.1 | ||||
AK293677.1 | ||||
AK298157.1 | ||||
BC000704.2 | AAH00704.1 | |||
BC004280.1 | AAH04280.1 | |||
BC009248.2 | AAH09248.1 | |||
BC011206.1 | AAH11206.1 | |||
DA359645.1 | ||||
DB449004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517068 | Chr.15:76962203 - 77088880 on Build GRCh38 | Gain+Loss | TSPAN3 PSTPIP1 |
esv2760041 | Chr.15:76878209 - 77081106 on Build GRCh38 | Gain | TSPAN3 PSTPIP1 SCAPER RCN2 |
nsv833058 | Chr.15:76943554 - 77101298 on Build GRCh38 | Loss | TSPAN3 PSTPIP1 RCN2 PEAK1 |
esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion | FBXO22-AS1 HMG20A MIR3713 LOC645752 LOC101929457 LINGO1 LINGO1-AS1 RCN2 PEAK1 ISL2 TSPAN3 PSTPIP1 LOC441728 FBXO22 ETFA TMEM266 UBE2Q2 LINC00597 SCAPER NRG4 TYRO3P LINGO1-AS2 LOC101929439 |
nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain | TSPAN3 PSTPIP1 HMG20A LINC00597 LOC101929457 SCAPER LINGO1-AS2 LINGO1 LINGO1-AS1 RCN2 PEAK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |