Assay Details
Target Gene Details
Entrez Gene ID: | 22801 |
Gene Name: | integrin subunit alpha 11 |
Gene Aliases: |
HsT18964 |
Location: |
Chr.15:68297433-68432312 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGA11 | NM_001004439.1 | NP_001004439.1 | ||
XM_005254228.3 | XP_005254285.1 | |||
XM_011521363.2 | XP_011519665.1 | |||
AF109681.1 | AAF01258.1 | |||
AF111799.1 | AAL39001.1 | |||
AF137378.2 | AAD51919.2 | |||
AK095863.1 | ||||
AK292445.1 | ||||
AK304778.1 | ||||
AL359064.1 | CAB94392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33024 | Chr.15:68299521 - 68336679 on Build GRCh38 | Gain | ITGA11 |
esv2760036 | Chr.15:68245734 - 68522324 on Build GRCh38 | Loss | ITGA11 FEM1B CORO2B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |