Assay Details
Target Gene Details
Entrez Gene ID: | 220 |
Gene Name: | aldehyde dehydrogenase 1 family member A3 |
Gene Aliases: |
ALDH1A6, ALDH6, MCOP8, RALDH3 |
Location: |
Chr.15:100879692-100916626 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH1A3 | NM_000693.3 | NP_000684.2 | ||
NM_001293815.1 | NP_001280744.1 | |||
AK302607.1 | ||||
AK303441.1 | ||||
AK312303.1 | ||||
AL110109.1 | ||||
BC069274.1 | AAH69274.1 | |||
BX538027.1 | CAD97973.1 | |||
U07919.1 | AAA79036.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041172 | Chr.15:100435008 - 101062634 on Build GRCh38 | Gain | LINS1 ASB7 LOC102723335 LRRK1 ALDH1A3 LOC105369201 LOC101927751 CERS3 PRKXP1 CERS3-AS1 LOC440313 |
nsv457288 | Chr.15:100884530 - 100924371 on Build GRCh38 | Loss | LRRK1 ALDH1A3 LOC101927751 |
nsv469824 | Chr.15:100759595 - 100934123 on Build GRCh38 | Loss | LRRK1 ALDH1A3 LOC105369201 LOC101927751 LOC440313 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |