Assay Details
Target Gene Details
Entrez Gene ID: | 57082 |
Gene Name: | kinetochore scaffold 1 |
Gene Aliases: |
AF15Q14, CASC5, CT29, D40, MCPH4, PPP1R55, Spc7, hKNL-1, hSpc105 |
Location: |
Chr.15:40594012-40664342 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KNL1 | NM_144508.4 | NP_653091.3 | ||
NM_170589.4 | NP_733468.3 | |||
XM_017022432.1 | XP_016877921.1 | |||
AB022190.1 | BAC05691.1 | |||
AF173994.1 | AAM45143.1 | |||
AF248041.1 | AAF97513.1 | |||
AF461041.1 | AAL67803.1 | |||
AK307605.1 | ||||
AK307996.1 | ||||
BC029373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2760026 | Chr.15:40540346 - 40702913 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv832984 | Chr.15:40502192 - 40706337 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)