Assay Details
Target Gene Details
Entrez Gene ID: | 101928388 |
Gene Name: | PLA2G4E antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:41972763-41999094 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G4E-AS1 | NR_120334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain | MAPKBP1 LOC105370792 SPTBN5 JMJD7-PLA2G4B PLA2G4D JMJD7 EHD4-AS1 PLA2G4F PLA2G4E PLA2G4B PLA2G4E-AS1 MIR4310 EHD4 |
nsv1038933 | Chr.15:41846258 - 42016889 on Build GRCh38 | Gain | EHD4-AS1 PLA2G4E PLA2G4B LOC105370792 SPTBN5 JMJD7-PLA2G4B PLA2G4E-AS1 MIR4310 EHD4 |